missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid Solution (0.02N (N/50)/Certified), Fisher Chemical™

Standardized to pH 4.4 at 25°C against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate

Supplier:  Thermo Fisher Scientific SA22620

 View more versions of this product

Catalog No. SA226-20


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4.4
Colorless
7664-93-9,7732-18-5,50-00-0
0.0198 to 0.0202N
PolyPac™
20 L
Liquid
2796
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
Certified
Sulfuric Acid Solution
0.02N (N/50)
100°C
Pass Test
LOT 84L Potassium acid phthalate
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
OS(O)(=O)=O
98.07
CHEBI:26836
0.02N (N/50)

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
Contains a known or suspected carcinogen. The toxicological properties have not been fully investigated. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:0
Instability:0

DOTInformation : DOT Class 8, : Corrosive

SDS Documents

Documents

Promotions

Promotions