missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid, Puriss. p.a., for determination of Hg, ACS Reagent, Reag. ISO, Reag. Ph. Eur., 95.0-97.0%, Honeywell Fluka™
Puriss. p.a., for determination of Hg, ACS Reagent, Reag. ISO, Reag. Ph. Eur., 95.0-97.0%
$83.23 - $1574.30
Chemical Identifiers
| CAS | 7664-93-9 |
|---|---|
| Molecular Formula | H2O4S |
| Molecular Weight (g/mol) | 98.07 |
| MDL Number | MFCD00064589 |
| InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
| Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
| PubChem CID | 1118 |
| ChEBI | CHEBI:26836 |
| IUPAC Name | sulfuric acid |
| SMILES | OS(O)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
6001593
|
Honeywell Inc
307431LGL |
1 L | Glass Bottle |
Each for $83.23
|
|
||||
|
6001592
|
Honeywell-Fluka
307431L |
1 L | Plastic Bottle |
Each for $196.73
|
|
||||
|
6001594
|
Honeywell Inc
3074325L |
2.5 L | Plastic Bottle |
Each for $310.10
|
|
||||
|
6001596
|
Honeywell-Fluka
307435L |
5 L | Plastic Bottle |
Each for $452.65
|
|
||||
|
60045850
|
Honeywell-Fluka
307436X1L |
6 x 1 L | Plastic Bottle |
Case of 6 Each for $1,038.40
|
|
||||
|
60045848
|
Honeywell-Fluka
307434X25LGL |
4 x 2.5 L | Glass Bottle |
Case of 4 Each for $633.48
|
|
||||
|
60045847
|
Honeywell-Fluka
307434X25L |
4 x 2.5 L | Plastic Bottle |
Case of 4 Each for $1,092.18
|
|
||||
|
60045849
|
Honeywell Inc
307434X5L |
4 x 5 L | Plastic Bottle |
Case of 4 Each for $1,574.30
|
|
||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
| 7664-93-9 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| 1118 | |
| sulfuric acid |
| H2O4S | |
| MFCD00064589 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| CHEBI:26836 | |
| OS(O)(=O)=O |
Specifications
| 7664-93-9 | |
| Glass Bottle | |
| 1mmHg (146°C) | |
| H2O4S | |
| MFCD00064589 | |
| 2037554 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| 98.08g/mol | |
| ACS/puriss. p.a. |
| 1.840g/mL at 25°C | |
| ≤5ppm | |
| 1 L | |
| H2SO4 | |
| 1830 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| OS(O)(=O)=O | |
| 98.07 | |
| CHEBI:26836 | |
| 95 to 97% | |
| Sulfuric Acid, Reag. Ph. Eur. reag. ISO |
Safety and Handling
P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
H290-H314
EINECSNumber : 231-639-5
RTECSNumber : WS5600000