missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid, Puriss. p.a., for determination of Hg, ACS Reagent, Reag. ISO, Reag. Ph. Eur., 95.0-97.0%, Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Density | 1.840g/mL at 25°C |
| Packaging | Plastic Bottle |
| Residue after Ignition | ≤5ppm |
| Vapor Pressure | 1mmHg (146°C) |
| Quantity | 2.5 L |
| Linear Formula | H2SO4 |
| UN Number | 1830 |
| Beilstein | 2037554 |
| Formula Weight | 98.08g/mol |
| Percent Purity | 95 to 97% |
| Show More |
Safety and Handling
| Hazard Statement | P260-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338 |
| Precautionary Statement | H290-H314 |
| EINECSNumber | 231-639-5 |
| RTECSNumber | WS5600000 |