missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric acid, Puriss., meets analytical spec. of BP, Ph. Eur., 95-97%, Honeywell Fluka™
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Boiling Point | 310°C |
| CAS Min % | ≥50.0000%, ≥1.0000% |
| Density | 1.840 g/cm3 |
| CAS Max % | ≤100.0000%, <5.0000% |
| Packaging | Plastic Bottle |
| Quantity | 4 x 5 L |
| Linear Formula | H2SO4 |
| UN Number | UN1830 |
| Beilstein | 2037554 |
| Formula Weight | 98.08 |
| Show More |
Safety and Handling
| ShelfLife | 1080 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Sulfuric acid |
| EINECSNumber | 231-639-5 |
| RTECSNumber | WS5600000 |
| Recommended Storage | Room Temp |