Learn More
Sulfuric Acid, FCC, 95-98%, Spectrum™ Chemical
S1736, 7664-93-9, H2SO4
$460.43 - $3,024.70
Chemical Identifiers
| CAS | 7664-93-9 |
|---|---|
| Molecular Formula | H2O4S |
| Molecular Weight (g/mol) | 98.07 |
| MDL Number | MFCD00064589 |
| InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
| IUPAC Name | sulfuric acid |
| SMILES | OS(O)(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18607165
|
Spectrum Chemical Mfg Cor
S1736500MLGL |
500 mL | Amber Glass Bottle |
N/A
|
|
||||
|
18607162
|
Spectrum Chemical Mfg Cor
S173625LTGL |
2.5 L | Glass Bottle |
N/A
|
|
||||
|
18607161
|
Spectrum Chemical Mfg Cor
S173625LTPV |
2.5 L | Poly-Coated Glass Bottle |
N/A
|
|
||||
|
18607166
|
Spectrum Chemical Mfg Cor
S1736500MLGLCS6 |
6 x 500 mL | Amber Glass Bottle |
N/A
|
|
||||
|
18607164
|
Spectrum Chemical Mfg Cor
S173625LTGLCS4 |
4 x 2.5 L | Glass Bottle |
N/A
|
|
||||
|
18607163
|
Spectrum Chemical Mfg Cor
S173625LTPVCS4 |
4 x 2.5 L | Poly-Coated Glass Bottle |
N/A
|
|
||||
|
18607167
|
Spectrum Chemical Mfg Cor
S1736200LTBL |
200 L | Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Sulfuric Acid, FCC is considered a general purpose food additive and is used to directly control pH during the processing of food by discouraging the growth of bacteria and spoilage microbes. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.