missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, FCC, 95-98%, Spectrum™ Chemical

S1736, 7664-93-9, H2SO4

Supplier:  Spectrum Chemical Mfg Cor S173625LTGL

 View more versions of this product

Catalog No. 18607162


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Sulfuric Acid, FCC is considered a general purpose food additive and is used to directly control pH during the processing of food by discouraging the growth of bacteria and spoilage microbes. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

Sulfuric Acid
1
2.5 L
MFCD00064589
OS(O)(=O)=O
98.07
FCC
7664-93-9
Glass Bottle
H2O4S
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
95 to 98%

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
OS(O)(=O)=O
H2O4S
MFCD00064589
sulfuric acid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions