missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, Dilute R (98 g/L H2SO4), Ricca Chemical

Catalog No. p-3252532
Change view
Click to view available options
Quantity:
250 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
81408 250 mL Natural Poly Bottle
814016 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 81408 Supplier Ricca Chemical Company Supplier No. 81408
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
Concentration 98g/L H2SO4
CAS Min % 89.0, 9.01
CAS Max % 93.0, 9.37
Packaging Natural Poly Bottle
Quantity 250 mL
Physical Form Liquid
Grade Laboratory
Chemical Name or Material Sulfuric Acid, Dilute Reagent