missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, Dilute R (98 g/L H2SO4), Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| Concentration | 98g/L H2SO4 |
| CAS Min % | 89.0, 9.01 |
| CAS Max % | 93.0, 9.37 |
| Packaging | Natural Poly Bottle |
| Quantity | 250 mL |
| Physical Form | Liquid |
| Grade | Laboratory |
| Chemical Name or Material | Sulfuric Acid, Dilute Reagent |