missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acide sulfurique, certifié, 25,0% (v/v) ±0,5%, LabChem™
Spécifications
Spécifications
| Nom ou substance chimique | Sulfuric Acid |
| Couleur | Incolore |
| CAS % maximal | 64.95, 35.05 |
| Densité | 1.26g/mL |
| Désignation | Test de réussite |
| Emballage | Bouteille en polyéthylène |
| Gravité spécifique | 1.26g/mL |
| Traçabilité NIST | Tracable à NIST |
| Renseignements sur la décomposition | Composés soufrés; La décomposition thermique génère : Des vapeurs corrosives |
| Quantité | 1 L |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. Harmful to aquatic life. |
| Precautionary Statement | GHS P Statement Do not breathe mist, vapors, spray. Wear protective gloves, protective clothing, eye protection, face protection. Wash exposed skin thoroughly after handling. Avoid release to the environment. If swallowed: Rinse mouth. Do not induce vomiting. Immediately call a poison center/doctor. If on skin (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a poison center/doctor. Wash contaminated clothing before reuse. If inhaled: Remove person to fresh air and keep comfortable for breathing. Immediately call a poison center/doctor. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container to comply with local, state and federal regulations. |
| Recommended Storage | Température ambiante |