missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, Certified, 0.200N ±0.002N (0.1M), LabChem™

Catalog No. p-5388880
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Catalog No. Quantity
LC257001 500 mL
LC257002 1 L
LC257004 4 L
LC257005 20 L
4 options

Catalog No. LC257001

Supplier: LabChem LC257001

Only null left

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
Concentration 0.200N ±0.002N (0.1M)
CAS Max % 99, 1
Density 1.01g/mL
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.01g/mL
Traceability to NIST Traceable to NIST
Decomposition Information Sulfur compounds; Thermal decomposition generates: Corrosive vapors
Quantity 500 mL
Physical Form Liquid
Linear Formula H2SO4
UN Number UN2796
Solubility Information Soluble in water
Formula Weight 98.08
Grade Certified
Chemical Name or Material Sulfuric Acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature