missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, Certified, 0.0500N ±0.0005N (0.025M), LabChem™
Specifications
Specifications
| Color | Colorless |
| Concentration | 0.0500N ±0.0005N (0.025M) |
| CAS Max % | 99.75, 0.25 |
| Density | 1g/mL |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Specific Gravity | 1g/mL |
| Traceability to NIST | Traceable to NIST |
| Decomposition Information | Sulfur compounds |
| Quantity | 1 L |
| Show More |
Safety and Handling
| missing translation for 'healthHazard2' | GHS H Statement Solution is not hazardous. |
| missing translation for 'healthHazard3' | GHS P Statement Wear eye protection, protective gloves. Wash exposed skin thoroughly after handling. If on skin: Wash with plenty of soap and water. Take off contaminated clothing. If skin irritation occurs: Get medical advice/attention. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. If eye irritation persists: Get medical advice/attention. |
| Recommended Storage | Room Temperature |