missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, Certified, 0.0100N ±0.0001N (0.005M), LabChem™

Catalog No. p-5388871
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Catalog No. Quantity
LC256551 500 mL
LC256552 1 L
LC256554 4 L
LC256555 20 L
4 options

Catalog No. LC256551

Supplier: LabChem LC256551

Only null left

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
Concentration 0.0100N ±0.0001N (0.005M)
CAS Max % 99.95, 0.05
Density 1g/mL
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Decomposition Information Sulfur compounds
Quantity 500 mL
Viscosity 1cSt
Physical Form Liquid
Linear Formula H2SO4
Solubility Information Soluble in water
Formula Weight 98.08
Grade Certified
Chemical Name or Material Sulfuric Acid
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature