missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid for Babcock Test, Certified, 90.5% (w/w) to 93.0% (w/w), LabChem™

Catalog No. p-5388869
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
2.5 L
Catalog No. Quantity
LC256502 1 L
LC256503 2.5 L
2 options

Catalog No. LC256502

Supplier: LabChem LC256502

Only null left

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
CAS Max % 92, 8
Density 1.8g/mL
Identification Passes Test
Packaging Amber Glass
Specific Gravity 1.8g/mL
Traceability to NIST Traceable to NIST
Decomposition Information Fume; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
Quantity 1 L
Physical Form Liquid
Linear Formula H2SO4
UN Number UN1830
Solubility Information Soluble in water
Formula Weight 98.08
Percent Purity 90.5 to 93.0% (w/w)
Grade Certified
Chemical Name or Material Sulfuric Acid for Babcock Test
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature