missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid for Babcock Test, Certified, 90.5% (w/w) to 93.0% (w/w), LabChem™

Supplier:  LabChem LC256503

 View more versions of this product

Catalog No. LC256503

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sulfuric Acid for Babcock Test
7664-93-9,7732-18-5
92,8
HDPE Bottle
Traceable to NIST
2.5 L
Liquid
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
98.08
Certified
Colorless
1.8g/mL
Passes Test
1.8g/mL
Fume; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
H2O4S
H2SO4
UN1830
Soluble in water
OS(O)(=O)=O
98.07
CHEBI:26836
90.5 to 93.0% (w/w)

Chemical Identifiers

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions