missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, Babcock, specific gravity 1.825, Ricca Chemical

Catalog No. 819216
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Safety-coated Amber Glass Bottle
Safety-coated Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
819216 500 mL Safety-coated Glass Bottle
819232 1 L Safety-coated Glass Bottle
81925PT 2.5 L Safety-coated Glass Bottle
81921 4 L Safety-coated Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 819216 Supplier Ricca Chemical Company Supplier No. 819216
Only null left

APHA for Fat, Babcock Method

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
CAS Min % 94.0, 4.31
CAS Max % 98.0, 4.49
Packaging Safety-coated Glass Bottle
Quantity 500 mL
Physical Form Liquid
Grade Laboratory
Chemical Name or Material Sulfuric Acid, specific gravity 1.825