missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, ACS reagent, 95% solution in water

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

Supplier:  Thermo Scientific Chemicals 424520100

 Afficher plus de versions de ce produit

Numéro de catalogue. AC424520100


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

Sulfuric acid
7664-93-9
1.8300g/mL
Plastic drum
1.83
10 L
H2O4S
H2SO4
01,470; 05,633; 06,558; 07,347; 09,441
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
QAOWNCQODCNURD-UHFFFAOYSA-N
sulfuric acid
1118
98.08
ACS Reagent
10.0°C
290.0°C
1ppm max.
0.0005% max.
95%
21 mPa.s (25°C)
Liquid Free From Suspended or Insoluble Matter
MFCD00064589
15, 9104
Solubility in water: miscible. Other solubilities: miscible with alcohol (generation of heat)
OS(O)(=O)=O
98.07
CHEBI:26836
95.0 to 98.0%

Identifiants chimiques

7664-93-9
98.07
QAOWNCQODCNURD-UHFFFAOYSA-N
1118
sulfuric acid
H2O4S
MFCD00064589
oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
CHEBI:26836
OS(O)(=O)=O
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

FDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only