missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric acid, ACS reagent, 95% solution in water

Catalog No. p-4228972
Change view
Click to view available options
Quantity:
500 mL
2.5 L
10 L
Packaging:
Glass Bottle
HDPE Bottles
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC424525000 500 mL Glass Bottle
AC424525001 500 mL Plastic bottle
AC424520025 2.5 L HDPE Bottles
AC424520100 10 L Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC424525000 Supplier Thermo Scientific Chemicals Supplier No. 424525000
Only null left

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Melting Point 10.0°C
Boiling Point 290.0°C
Density 1.8300g/mL
Heavy Metals (as Pb) 1ppm max.
Packaging Glass Bottle
Residue after Ignition 0.0005% max.
Specific Gravity 1.83
Assay Percent Range 95%
Quantity 500 mL
Viscosity 21 mPa.s (25°C)
Physical Form Liquid Free From Suspended or Insoluble Matter
Linear Formula H2SO4
Fieser 01,470; 05,633; 06,558; 07,347; 09,441
Merck Index 15, 9104
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol (generation of heat)
Formula Weight 98.08
Percent Purity 95.0 to 98.0%
Grade ACS Reagent
Chemical Name or Material Sulfuric acid
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only