missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, ACS Reagent, Reagents

Catalog No. 01336807
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336807 1 L
01336806 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336807 Supplier Labchem Inc Supplier No. C51026401A
Only null left

Sulfuric Acid, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym Oil of Vitriol, Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 0
Color Colorless
Boiling Point 330°C
Packaging Poly Bottle
Specific Gravity 1.84
Quantity 1 L
Physical Form Liquid
Grade ACS
Chemical Name or Material Sulfuric Acid, ACS Reagent