missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, ACS Reagent, Reagents

Catalog No. p-200021100
Change view
Click to view available options
Quantity:
1 L
2.5 L
75 lb.
750 lb.
Packaging:
Glass Bottle
HDPE Winpak
Poly Drum
Safety Coated Glass
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01336812 1 L Glass Bottle
01336811 2.5 L Safety Coated Glass
01336810 2.5 L Glass Bottle
01336809 750 lb. Poly Drum
01336808 75 lb. HDPE Winpak
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01336812 Supplier Labchem Inc Supplier No. C51026001C
Only null left

Sulfuric Acid, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym Oil of Vitriol, Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 0
Color Colorless
Boiling Point 330°C
Packaging Glass Bottle
Specific Gravity 1.84
Quantity 1 L
Physical Form Liquid
Grade ACS
Chemical Name or Material Sulfuric Acid, ACS Reagent