missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, ACS Reagent Grade, Ricca Chemical
Supplier: Ricca Chemical Company RABS003025D
Specifications
| Sulfuric Acid | |
| 7664-93-9 | |
| 98.0 | |
| Safety-coated Glass Bottle | |
| 2.5 L | |
| Liquid | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| OS(O)(=O)=O | |
| 98.07 | |
| CHEBI:26836 | |
| ACS |
| Colorless | |
| 95.0 | |
| max. 0.001% | |
| Pass | |
| H2O4S | |
| MFCD00064589 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| ≥85% |
Chemical Identifiers
| 7664-93-9 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| CHEBI:26836 | |
| OS(O)(=O)=O |
| H2O4S | |
| MFCD00064589 | |
| 1118 | |
| sulfuric acid |
Safety and Handling
ShelfLife : 36 months