missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, ACS Reagent Grade, Ricca Chemical

Catalog No. p-3252530
Change view
Click to view available options
Quantity:
500 mL
2.5 L
4 L
Packaging:
Amber Glass Bottle
Natural Poly Bottle
Safety-coated Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
RABS0030500 500 mL Amber Glass Bottle
RABS003025D 2.5 L Safety-coated Glass Bottle
RABS00304A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. RABS0030500 Supplier Ricca Chemical Company Supplier No. RABS0030500C
Only null left

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
CAS Min % 95.0
CAS Max % 98.0
Heavy Metals (as Pb) max. 0.001%
Packaging Amber Glass Bottle
Reducing Substances Pass
Quantity 500 mL
Physical Form Liquid
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
Percent Purity ≥85%
Grade ACS
Chemical Name or Material Sulfuric Acid
Show More Show Less
ShelfLife 36 months