Learn More
Sulfuric Acid, ACS Grade, 95.0 to 98.0% (w/w), LabChem™
Supplier: LabChem LC255503S
Specifications
| Sulfuric Acid | |
| 10°C | |
| 288°C | |
| 100 | |
| PVC Coated | |
| 1840kg/m3 | |
| 2.5 L | |
| Liquid | |
| MFCD00064589 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1118 | |
| 98.08 | |
| ACS |
| Colorless | |
| 7664-93-9 | |
| 1,840kg/m3 | |
| Passes Test | |
| ≤2ppm | |
| Sulfur compounds | |
| H2O4S | |
| H2SO4 | |
| UN1830 | |
| Soluble in water | |
| OS(O)(=O)=O | |
| 98.07 | |
| CHEBI:26836 | |
| 95.0 to 98.0% (w/w) |
Chemical Identifiers
| 7664-93-9 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| 1118 | |
| sulfuric acid |
| H2O4S | |
| MFCD00064589 | |
| oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil | |
| CHEBI:26836 | |
| OS(O)(=O)=O |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Danger
EINECSNumber : 231-639-5
Recommended Storage : Room Temperature