missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, ACS Grade, 95.0 to 98.0% (w/w), LabChem™

Numéro de catalogue. p-5379570
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 2.5 L
Packaging:
Amber Glass
HDPE Bottle
PVC Coated
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
LC255501 500 mL Amber Glass
LC255502 1 L Amber Glass
LC255503 2.5 L HDPE Bottle
LC255503S 2.5 L PVC Coated
LC2555036 6 x 2.5 L HDPE Bottle
LC2555036S 6 x 2.5 L PVC Coated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Numéro de catalogue. LC255501 Fournisseur LabChem Code fournisseur LC255501
Il en reste null

Identifiants chimiques

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Spécifications

Color Colorless
Melting Point 10°C
Boiling Point 288°C
CAS Max % 100
Density 1,840kg/m3
Identification Passes Test
Packaging Amber Glass
Reducing Substances ≤2ppm
Specific Gravity 1840kg/m3
Decomposition Information Sulfur compounds
Quantity 500 mL
Physical Form Liquid
Linear Formula H2SO4
UN Number UN1830
Solubility Information Soluble in water
Formula Weight 98.08
Percent Purity 95.0 to 98.0% (w/w)
Grade ACS
Chemical Name or Material Sulfuric Acid
Afficher plus de résultats Afficher moins de résultats
Hazard Category Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 231-639-5
Recommended Storage Room Temperature