missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, ACS, 95.0-98.0%, Spectrum™ Chemical

$230.00 - $9904.37

Chemical Identifiers

CAS 7664-93-9
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18614293
View Documents
Spectrum Chemical Mfg Cor
S1730500MLGL
500 mL Amber Glass Bottle
Each for $230.00
Only null left
 
18614291
View Documents
Spectrum Chemical Mfg Cor
S173025LTGL
2.5 L Glass Bottle
Each for $447.73
Only null left
 
18614290
View Documents
Spectrum Chemical Mfg Cor
S173025LTPV
2.5 L Poly-Coated Glass Bottle
Each for $489.83
Only null left
 
18614289
View Documents
Spectrum Chemical Mfg Cor
S1730185LTBL
185 L Drum
Each for $9,904.37
Only null left
 
18614292
View Documents
Spectrum Chemical Mfg Cor
S17305GLBL
5 gal. Drum
Each for $2,573.70
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7664-93-9

Specifications

7664-93-9
1 ppm
5 ppm
95 to 98%
Sulfuric Acid
1
Amber Glass Bottle
500 mL
Reagent
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.