missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, ACS, 95.0-98.0%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S173025LTPV

 View more versions of this product

Catalog No. 18614290


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sulfuric Acid
1
Poly-Coated Glass Bottle
2.5 L
Reagent
7664-93-9
1 ppm
5 ppm
95 to 98%

Chemical Identifiers

7664-93-9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.