missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 96%, Suprapur™, MilliporeSigma™
Supplier: Technidata 1007140250
Specifications
| 0.3 (49Â g/L, H2O, 25°C) | |
| 7664-93-9 | |
| Glass Bottle | |
| 250 mL | |
| MFCD00064589 | |
| OS(O)(=O)=O | |
| 98.07 |
| −20°C | |
| 1.84Â g/cm3 (at 20°C) | |
| 0.0001Â hPa (20°C) | |
| H2O4S | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid |
Chemical Identifiers
| 7664-93-9 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| OS(O)(=O)=O |
| H2O4S | |
| MFCD00064589 | |
| sulfuric acid |