missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 96%, Suprapur™, MilliporeSigma™

Catalog No. p-7162461
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1007140250 250 mL
M1007141000 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1007140250 Supplier Technidata Supplier No. 1.00714.0250
Only null left

Chemical Identifiers

CAS 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH 0.3 (49 g/L, H2O, 25°C)
Melting Point −20°C
Density 1.84 g/cm3 (at 20°C)
Packaging Glass Bottle
Vapor Pressure 0.0001 hPa (20°C)
Quantity 250 mL