missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 72% (w/w), Ricca Chemical

Catalog No. R81916001A
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 L
Packaging:
Natural Poly Bottle
Safety-coated Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R81916001A 1 L Natural Poly Bottle
R819160025D 2.5 L Safety-coated Glass Bottle
R81916004A 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R81916001A Supplier Ricca Chemical Company Supplier No. R81916001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
Concentration 72% (w/w)
CAS Min % 71.0, 27.0
CAS Max % 73.0, 29.0
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 1 L
Physical Form Liquid
Percent Purity 72% W/W
Grade Laboratory
Chemical Name or Material Sulfuric Acid
Show More Show Less