missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 6.0 Normal, Reagents

Sulfuric Acid, 6.0 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

$78.77 - $557.63

Chemical Identifiers

CAS 7732-18-5
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name water
SMILES O
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
01-333-822
View Documents
Labchem Inc
CS1061001A
1 L
Each for $98.60
Only null left
 
01-333-821
View Documents
Labchem Inc
CS10610020F
20 L
Each for $557.63
Only null left
 
01-333-820
View Documents
Labchem Inc
CS1061004A
4 L
Each for $302.17
Only null left
 
01-333-819
View Documents
Labchem Inc
CS106100500A
500 mL
Each for $78.77
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5
18.02
Sulphuric Acid, Dihydrogen Sulfate
O
H2O
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water

Specifications

<1
7732-18-5,7664-93-9
Poly Bottle
1 L
Liquid
XLYOFNOQVPJJNP-UHFFFAOYSA-N
water
Reagent
Colorless
105°C
1.184
H2O
Sulphuric Acid, Dihydrogen Sulfate
O
18.02
Sulfuric Acid, 6.0 Normal
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'