missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 50% (w/w) Solution, Spectrum™ Chemical

$144.16 - $535.06

Chemical Identifiers

CAS 7664-93-9
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18614278
View Documents
Spectrum Chemical Mfg Cor
S311500ML
500 mL Amber Glass Bottle N/A
Only null left
 
18614277
View Documents
Spectrum Chemical Mfg Cor
S31125LTGL
2.5 L Glass Bottle N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7664-93-9

Specifications

7664-93-9,7732-18-5
Amber Glass Bottle
50% ±1% w/w
Sulfuric Acid
50%
500 mL
Ungraded
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.