missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 50% (w/w) Solution, Spectrum™ Chemical
Specifications
Specifications
| Concentration | 50% |
| Packaging | Amber Glass Bottle |
| Quantity | 500 mL |
| Percent Purity | 50% ±1% w/w |
| Grade | Ungraded |
| Chemical Name or Material | Sulfuric Acid |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.