missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 5.00 N, Ricca Chemical
Specifications
Specifications
| Color | Colorless |
| Concentration | 5.00Normal |
| CAS Min % | 78.0, 19.96 |
| CAS Max % | 81.0, 20.77 |
| Packaging | Natural Poly Bottle |
| Quantity | 1 L |
| Physical Form | Liquid |
| Grade | Laboratory |
| Chemical Name or Material | Sulfuric Acid |