missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 5.0 Normal, Reagents
Sulfuric Acid, 5.0 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.
$133.10 - $811.20
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | Sulphuric Acid, Dihydrogen Sulfate |
| IUPAC Name | water |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-333-625
|
Labchem Inc
CS14550010F |
10 L |
N/A
|
|
|||||
|
01-333-624
|
Labchem Inc
CS1455001A |
1 L |
N/A
|
|
|||||
|
01-333-623
|
Labchem Inc
CS14550020F |
20 L |
N/A
|
|
|||||
|
01-333-622
|
Labchem Inc
CS1455004A |
4 L |
N/A
|
|
|||||
|
01-333-621
|
Labchem Inc
CS145500500A |
500 mL |
N/A
|
|
|||||
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Sulphuric Acid, Dihydrogen Sulfate | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
Specifications
| <1 | |
| 7732-18-5,7664-93-9 | |
| Cubitainer | |
| 10 L | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent |
| Colorless | |
| 105°C | |
| 1.15 | |
| H2O | |
| Sulphuric Acid, Dihydrogen Sulfate | |
| O | |
| 18.02 | |
| Sulfuric Acid, 5.0 Normal |