missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 4.0 Normal, Reagents

Catalog No. p-200021699
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Fortpack
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01336330 10 L Cubitainer
01336329 1 L Poly Bottle
01336328 20 L Fortpack
01336327 20 L Cubitainer
01336326 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01336330 Supplier Labchem Inc Supplier No. C730590010F
Only null left

Sulfuric Acid, 4.0 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless
Boiling Point 105°C
Packaging Cubitainer
Specific Gravity 1.12
Quantity 10 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Sulfuric Acid, 4.0 Normal