missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 4.0 Normal, Reagents

Catalog No. 01336330
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Fortpack
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01336330 10 L Cubitainer
01336329 1 L Poly Bottle
01336328 20 L Fortpack
01336327 20 L Cubitainer
01336326 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01336330 Supplier Labchem Inc Supplier No. C730590010F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sulfuric Acid, 4.0 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless
Boiling Point 105°C
Packaging Cubitainer
Specific Gravity 1.12
Quantity 10 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Sulfuric Acid, 4.0 Normal