missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 3.50 N, Ricca Chemical

Catalog No. 8321351
Change view
Click to view available options
Quantity:
4 L
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
8321351 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 8321351 Supplier Ricca Chemical Company Supplier No. 8321.351
Only null left

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
Concentration 3.5Normal
Packaging Natural Poly Bottle
Quantity 4 L
Physical Form Liquid
Grade Laboratory
Chemical Name or Material Sulfuric Acid