missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 3.0 Normal, Reagents

Catalog No. 01333534
Change view
Click to view available options
Quantity:
1 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333534 20 L
01333535 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01333534 Supplier Labchem Inc Supplier No. CS20310020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sulfuric Acid, 3.0 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name water
SMILES O

Specifications

pH 0
Color Colorless
Boiling Point 105°C
Packaging Cubitainer
Specific Gravity 1.094
Quantity 20 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Sulfuric Acid, 3.0 Normal