missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 25% (v/v), Ricca Chemical

Catalog No. p-2421874
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Safety-coated Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
817032 1 L Natural Poly Bottle
81705PT 2.5 L Safety-coated Glass Bottle
81701 4 L Natural Poly Bottle
81705 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 817032 Supplier Ricca Chemical Company Supplier No. 817032
Only null left

Meets EPA Specifications

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

CAS Min % 74.0, 24.0
CAS Max % 76.0, 26.0
Packaging Natural Poly Bottle
Quantity 1 L
Percent Purity 25% V/V
Grade Laboratory
Chemical Name or Material Sulfuric Acid, 25% v/v Aqueous, Ricca Chemical
Recommended Storage 15-30°C