missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 20% (v/v), Reagent Grade, Ricca Chemical

Catalog No. 816516
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
816516 500 mL Natural Poly Bottle
816532 1 L Natural Poly Bottle
81651 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 816516 Supplier Ricca Chemical Company Supplier No. 816516

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Appearance of Solution Colorless Liquid
Concentration 20% (v/v)
CAS Min % 79.0, 19.0
CAS Max % 81.0, 21.0
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Quantity 500 mL
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
Percent Purity 19.97 to 20.03%
Grade Reagent
Chemical Name or Material Sulfuric Acid, 20% (v/v)
Show More Show Less
ShelfLife 36 months