missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 2.00 N, Ricca Chemical

Catalog No. 831016
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
Safety-coated Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
831016 500 mL Natural Poly Bottle
831032 1 L Natural Poly Bottle
83105PT 2.5 L Safety-coated Glass Bottle
R83100004F 4 L Cubitainer
83101 4 L Natural Poly Bottle
83105 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 831016 Supplier Ricca Chemical Company Supplier No. RI831016

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
Concentration 2.00Normal
CAS Min % 89.0, 9.11
CAS Max % 93.0, 9.48
Packaging Natural Poly Bottle
Quantity 500 mL
Physical Form Liquid
Grade Laboratory
Chemical Name or Material Sulfuric Acid