missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 19.2 Normal, Reagents

Catalog No. 01333619
Change view
Click to view available options
Quantity:
500 mL
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01333619 4 L
01333618 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01333619 Supplier Labchem Inc Supplier No. CS1458004A
Only null left

Sulfuric Acid, 19.2 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
Synonym Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

pH <1
Color Colorless
Boiling Point 107°C
Packaging Poly Bottle
Specific Gravity 1.5223
Quantity 4 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Sulfuric Acid, 19.2 Normal