missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 18.0 N, Ricca Chemical

Catalog No. 83431832
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
83431832 1 L Natural Poly Bottle
83431816 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 83431832 Supplier Ricca Chemical Company Supplier No. 8343.1832

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7664-93-9 | H2O4S | 98.07 g/mol

Chemical Identifiers

CAS 7664-93-9 , 7732-18-5
Molecular Formula H2O4S
Molecular Weight (g/mol) 98.07
MDL Number MFCD00064589
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
PubChem CID 1118
ChEBI CHEBI:26836
IUPAC Name sulfuric acid
SMILES OS(O)(=O)=O

Specifications

Color Colorless
Appearance of Solution Colorless Liquid
Concentration 17.95 to 18.05 N
CAS Min % 51.0, 47.0
CAS Max % 53.0, 48.92
Packaging Natural Poly Bottle
Quantity 1 L
Physical Form Liquid
Synonym oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil
Grade Laboratory
Assay Normal at 20°C
Chemical Name or Material Sulfuric Acid
Show More Show Less
ShelfLife 36 months