missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 12 Normal, Reagents
Sulfuric Acid, 12 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.
Supplier: Labchem Inc CS1078004A
Specifications
| <1 | |
| Colorless | |
| 107°C | |
| 1.34 | |
| H2O | |
| Sulphuric Acid, Dihydrogen Sulfate | |
| O | |
| 18.02 |
| Sulfuric Acid, 12 Normal | |
| 7732-18-5,7664-93-9 | |
| Poly Bottle | |
| 4 L | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Sulphuric Acid, Dihydrogen Sulfate | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |