missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 1.00 Normal, Reagents
Sulfuric Acid, 1.00 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.
Supplier: Labchem Inc C73055604F
Specifications
| <1 | |
| Colorless | |
| 100°C | |
| 1.031 | |
| H2O | |
| Sulphuric Acid, Dihydrogen Sulfate | |
| O | |
| 18.02 |
| Sulfuric Acid, 1.00 Normal | |
| 7732-18-5,7664-93-9 | |
| Cubitainer | |
| 4 L | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Sulphuric Acid, Dihydrogen Sulfate | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |