missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 0.940 Normal, Reagents
Specifications
Specifications
| pH | <1 |
| Color | Colorless |
| Boiling Point | 100°C |
| Packaging | Poly Bottle |
| Specific Gravity | 1.03 |
| Quantity | 1 L |
| Physical Form | Liquid |
| Grade | Reagent |
| Chemical Name or Material | Sulfuric Acid, 0.940 Normal |