Learn More
Sulfuric acid, 0.5N Standardized Solution
CAS: 7664-93-9 | H2O4S | 98.07 g/mol
$91.63 - $321.29
Chemical Identifiers
| CAS | 7664-93-9 |
|---|---|
| Molecular Formula | H2O4S |
| Molecular Weight (g/mol) | 98.07 |
| MDL Number | MFCD00064589 |
| InChI Key | QAOWNCQODCNURD-UHFFFAOYSA-N |
| PubChem CID | 1118 |
| ChEBI | CHEBI:26836 |
| IUPAC Name | sulfuric acid |
| SMILES | OS(O)(=O)=O |
Description
Sulfuric acid, 0.5N standardized solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7664-93-9 | |
| 98.07 | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| CHEBI:26836 | |
| OS(O)(=O)=O |
| H2O4S | |
| MFCD00064589 | |
| 1118 | |
| sulfuric acid |
Specifications
| 7664-93-9,7732-18-5 | |
| Liquid | |
| MFCD00064589 | |
| Miscible with water | |
| OS(O)(=O)=O | |
| 98.07 | |
| 1118 | |
| 98.08 | |
| Sulfuric acid |
| 23 hPa (17mm Hg) at 20°C | |
| H2O4S | |
| Colorless to Yellow | |
| QAOWNCQODCNURD-UHFFFAOYSA-N | |
| sulfuric acid | |
| 1 L | |
| CHEBI:26836 | |
| Sulfuric acid: 5%; Water: 95% |
Safety and Handling
P234-P261-P264b-P271-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P332+P313-P362-P390-P501c
H290-H315-H318-H335
DOTInformation : DOT Class: 8, Packaging Group: III
EINECSNumber : 231-639-5
RTECSNumber : WS5600000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only