missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid, 0.255 Normal, Reagents
Sulfuric Acid, 0.255 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.
Supplier: Labchem Inc C73058004A
Specifications
| <1 | |
| Colorless | |
| 100°C | |
| 1.009 | |
| H2O | |
| Sulphuric Acid, Dihydrogen Sulfate | |
| O | |
| 18.02 |
| Sulfuric Acid, 0.255 Normal | |
| 7732-18-5,7664-93-9 | |
| Poly Bottle | |
| 4 L | |
| Liquid | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent |
Chemical Identifiers
| 7732-18-5 | |
| 18.02 | |
| Sulphuric Acid, Dihydrogen Sulfate | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |