missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 0.250 N, Reagents

Catalog No. p-200021390
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336351 1 L
01336350 20 L
01336349 4 L
01336348 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01336351 Supplier Labchem Inc Supplier No. C73055301A
Only null left

Sulfuric Acid, 0.250 N, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless
Boiling Point 100°C
Packaging Poly Bottle
Specific Gravity 1.0075
Quantity 1 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Sulfuric Acid, 0.250 N