missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 0.250 N, Reagents

Catalog No. 01336350
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336350 20 L
01336351 1 L
01336349 4 L
01336348 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01336350 Supplier Labchem Inc Supplier No. C730553020F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sulfuric Acid, 0.250 N, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name water
SMILES O

Specifications

pH <1
Color Colorless
Boiling Point 100°C
Packaging Cubitainer
Specific Gravity 1.0075
Quantity 20 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Sulfuric Acid, 0.250 N