missing translation for 'onlineSavingsMsg'
Learn More

Sulfuric Acid, 0.200 N, Reagents

Catalog No. 01336356
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336356 10 L
01336355 1 L
01336354 20 L
01336353 4 L
01336352 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01336356 Supplier Labchem Inc Supplier No. C730552010F

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Sulfuric Acid, 0.200 N, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7664-93-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sulphuric Acid, Dihydrogen Sulfate
IUPAC Name water
SMILES O

Specifications

pH 1
Color Colorless
Boiling Point 100°C
Packaging Cubitainer
Specific Gravity 1.0067
Quantity 10 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Sulfuric Acid, 0.200 N